Få mere at vide
5,6,7,8,9-Dehydro-10-desmethyl Finasteride, TRC
Artikelnummer. 30363785
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363785 1 mg
30362304 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30363785 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2295101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346602-40-1
Molekylær formel C22H28N2O2
Molekylvægt (g/mol) 352.47
Synonym 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-(1,1-dimethylethyl)-2,5,6,6a,7,8,9,9a-octahydro-6a-methyl-2-oxo-, (6aR,7R,9aS)-rel-, 5,6,7,8,9-Dehydro-10-desmethyl Finasteride, 5,6,7,8,9-Dehydro-10-desmethylfinasteride
SMIL C[C@]12CCC3=C([C@@H]1CC[C@@H]2C(=O)NC(C)(C)C)C=CC4=C3C=CC(=O)N4
Tekniske data
Kemisk navn eller materiale 5,6,7,8,9-Dehydro-10-desmethyl Finasteride
Mængde 1 mg
InChI formel InChI=1S/C22H28N2O2/c1-21(2,3)24-20(26)17-8-7-16-14-5-9-18-15(6-10-19(25)23-18)13(14)11-12-22(16,17)4/h5-6,9-10,16-17H,7-8,11-12H2,1-4H3,(H,23,25)(H,24,26)/t16-,17+,22-/m0/s1
Formel vægt 352.2151