Få mere at vide
5,5-Diphenyl-d10-hydantoin, CDN
Artikelnummer. 30314568
Skift visning
Mængde:
0.05 g
Artikelnummer. Mængde
30314568 0.05 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314568 Leverandør CDN Leverandørnr. CDND21580.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenytoin-D10
CAS 65854-97-9
Molekylær formel C15 2H10 H2 N2 O2
Mængde 0.05 g
Synonym 2,4-Imidazolidinedione, 5,5-di(phenyl-d5)- (9 CI), Phenytoin-D10, 5,5-(Diphenyl-d10)hydantoin
InChI formel InChI=1 S/C15H12N2O2/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10 H,(H2,16,17,18,19)/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C2(NC(=O)NC2=O)c3c([2 H])c([2 H])c([2 H])c([2 H])c3[2 H]
IUPAC navn 5,5-bis(2,3,4,5,6-pentadeuteriophenyl)imidazolidine-2,4-dione
Molekylvægt (g/mol) 262.33
Formel vægt 262.1526 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity