missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dibromobarbituric acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7077278
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
511-67-1
C4H2Br2N2O3
285.88
MFCD00010595
AMATXUCYHHHHHB-UHFFFAOYSA-N
5,5-dibromobarbituric acid, dibromin, 5,5-dibromopyrimidine-2,4,6 1h,3h,5h-trione, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dibromo, dibrombarbitursaure, dibromobarbituric acid, acmc-20am7j, barbituric acid,5-dibromo, diethyl trimethylsilylphosphate
94877
5,5-dibromo-1,3-diazinane-2,4,6-trione
BrC1(Br)C(=O)NC(=O)NC1=O
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
511-67-1
C4H2Br2N2O3
285.88
MFCD00010595
AMATXUCYHHHHHB-UHFFFAOYSA-N
5,5-dibromobarbituric acid, dibromin, 5,5-dibromopyrimidine-2,4,6 1h,3h,5h-trione, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dibromo, dibrombarbitursaure, dibromobarbituric acid, acmc-20am7j, barbituric acid,5-dibromo, diethyl trimethylsilylphosphate
94877
5,5-dibromo-1,3-diazinane-2,4,6-trione
BrC1(Br)C(=O)NC(=O)NC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 240.0°C to 242.0°C
Formula Weight 285.88
Quantity 1 g
Chemical Name or Material 5,5-Dibromobarbituric acid
compliance-icons
Product Identifier
  • 5,5-Dibromobarbituric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'