Få mere at vide
5-(5-Chloro-1-(2-fluorophenyl)-2-oxopentyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl Acetate Maleate, TRC
Artikelnummer. 30367775
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367775 5 mg
30361650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367775 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3667855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1373350-61-8
Molekylær formel C20 H21 Cl F N O3 S . C4 H4 O4
Molekylvægt (g/mol) 525.97
Synonym 2-Pentanone, 1-[2-(acetyloxy)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]-5-chloro-1-(2-fluorophenyl)-, (2Z)-2-butenedioate (1:1), 5-[(1RS)-5-Chloro-1-(2-fluorophenyl)-2-oxopentyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate maleate, 1-[2-(Acetyloxy)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]-5-chloro-1-(2-fluorophenyl)-2-pentanone maleate, 2-Acetoxy-5-(5-chloro-1-(2-fluorophenyl)-2-oxopentyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine maleate
IUPAC navn (Z)-but-2-enedioic acid;[5-[5-chloro-1-(2-fluorophenyl)-2-oxopentyl]-6,7-dihydro-4H-thieno[3,2-c]pyridin-2-yl] acetate
SMIL CC(=O)Oc1cc2CN(CCc2s1)C(C(=O)CCCCl)c3ccccc3F.OC(=O)\C=C/C(=O)O
Tekniske data
Kemisk navn eller materiale 5-(5-Chloro-1-(2-fluorophenyl)-2-oxopentyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate maleate
Mængde 5 mg
InChI formel InChI=1S/C20H21ClFNO3S.C4H4O4/c1-13(24)26-19-11-14-12-23(10-8-18(14)27-19)20(17(25)7-4-9-21)15-5-2-3-6-16(15)22;5-3(6)1-2-4(7)8/h2-3,5-6,11,20H,4,7-10,12H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formel vægt 525.1024