missing translation for 'onlineSavingsMsg'
Learn More

5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-boronic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7052600
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
169126-63-0
C14H21BO2
232.13
MFCD06801711
NXBNRLONOXGRCQ-UHFFFAOYSA-N
5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl boronic acid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene-2-boronic acid, 1,1,4,4-tetramethyltetralin-6-ylboronic acid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-ylboronic acid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthylboronic acid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-2-naphthalenylboronic acid, b-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl boronic acid, boronic acid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl
10353857
(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)boronic acid
B(C1=CC2=C(C=C1)C(CCC2(C)C)(C)C)(O)O
This item is not returnable. View return policy
169126-63-0
C14H21BO2
232.13
MFCD06801711
NXBNRLONOXGRCQ-UHFFFAOYSA-N
5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl boronic acid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene-2-boronic acid, 1,1,4,4-tetramethyltetralin-6-ylboronic acid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-ylboronic acid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthylboronic acid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-2-naphthalenylboronic acid, b-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl boronic acid, boronic acid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl
10353857
(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)boronic acid
B(C1=CC2=C(C=C1)C(CCC2(C)C)(C)C)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 195°C to 197°C
Color White
Quantity 1 g
Formula Weight 232.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'