missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Pyridyl)thieno[2,3-d]pyrimidin-4(3H)-one, 97%, Thermo Scientific™

Catalog Number 15424418
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1421263-78-6
C11H7N3OS
229.257
MFCD22988937
DUWHOSMEYMIDKJ-UHFFFAOYSA-N
5-4-pyridyl thieno 2,3-d pyrimidin-4 3h-one, 5-pyridin-4-yl-3h-thieno 2,3-d pyrimidin-4-one
68537372
5-pyridin-4-yl-3H-thieno[2,3-d]pyrimidin-4-one
C1=CN=CC=C1C2=CSC3=C2C(=O)NC=N3
This item is not returnable. View return policy
1421263-78-6
C11H7N3OS
229.257
MFCD22988937
DUWHOSMEYMIDKJ-UHFFFAOYSA-N
5-4-pyridyl thieno 2,3-d pyrimidin-4 3h-one, 5-pyridin-4-yl-3h-thieno 2,3-d pyrimidin-4-one
68537372
5-pyridin-4-yl-3H-thieno[2,3-d]pyrimidin-4-one
C1=CN=CC=C1C2=CSC3=C2C(=O)NC=N3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 229.25
Percent Purity 97%
Chemical Name or Material 5-(4-Pyridyl)thieno[2,3-d]pyrimidin-4(3H)-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'