missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-Pyridyl)thieno[2,3-d]pyrimidin-4(3H)-one, 97%, Alfa Aesar™
Brand: Alfa Aesar H58053.06
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
1421263-78-6 | |
229.257 | |
DUWHOSMEYMIDKJ-UHFFFAOYSA-N | |
68537372 | |
C1=CN=CC=C1C2=CSC3=C2C(=O)NC=N3 |
C11H7N3OS | |
MFCD22988937 | |
5-4-pyridyl thieno 2,3-d pyrimidin-4 3h-one, 5-pyridin-4-yl-3h-thieno 2,3-d pyrimidin-4-one | |
5-pyridin-4-yl-3H-thieno[2,3-d]pyrimidin-4-one |
Specifications
5-(4-Pyridyl)thieno[2,3-d]pyrimidin-4(3H)-one | |
1421263-78-6 | |
MFCD22988937 | |
DUWHOSMEYMIDKJ-UHFFFAOYSA-N | |
5-pyridin-4-yl-3H-thieno[2,3-d]pyrimidin-4-one | |
68537372 | |
97% |
5g | |
C11H7N3OS | |
5-4-pyridyl thieno 2,3-d pyrimidin-4 3h-one, 5-pyridin-4-yl-3h-thieno 2,3-d pyrimidin-4-one | |
C1=CN=CC=C1C2=CSC3=C2C(=O)NC=N3 | |
229.257 | |
229.25 |