missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Nitrophenyl)-2-furoic acid, Thermo Scientific™

Catalog Number p-3212180
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
28123-73-1
C11H6NO5
232.17
MFCD00094261
FXZTYDPWMQBTDH-UHFFFAOYSA-M
5-4-nitrophenyl furan-2-carboxylic acid, 5-4-nitrophenyl-2-furoic acid, 5-4-nitro-phenyl-furan-2-carboxylic acid, 2-furancarboxylic acid, 5-4-nitrophenyl, 2-furancarboxylic acid,5-4-nitrophenyl, 5-p-nitrophenyl-2-furancarboxylic acid, 5-4-nitrophenyl-2-furancarboxylic acid, 5-4-nitrophenyl-2-furan carboxylic acid
161593
5-(4-nitrophenyl)furan-2-carboxylate
[O-]C(=O)C1=CC=C(O1)C1=CC=C(C=C1)[N+]([O-])=O
This item is not returnable. View return policy
28123-73-1
C11H6NO5
232.17
MFCD00094261
FXZTYDPWMQBTDH-UHFFFAOYSA-M
5-4-nitrophenyl furan-2-carboxylic acid, 5-4-nitrophenyl-2-furoic acid, 5-4-nitro-phenyl-furan-2-carboxylic acid, 2-furancarboxylic acid, 5-4-nitrophenyl, 2-furancarboxylic acid,5-4-nitrophenyl, 5-p-nitrophenyl-2-furancarboxylic acid, 5-4-nitrophenyl-2-furancarboxylic acid, 5-4-nitrophenyl-2-furan carboxylic acid
161593
5-(4-nitrophenyl)furan-2-carboxylate
[O-]C(=O)C1=CC=C(O1)C1=CC=C(C=C1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 255°C to 257°C
Packaging Glass bottle
Quantity 1 g
Formula Weight 233.18
Chemical Name or Material 5-(4-Nitrophenyl)-2-furoic acid
compliance-icons
Product Identifier
  • 5-(4-Nitrophenyl)-2-furoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'