Få mere at vide
5-(4-Nitrophenyl)-2-furaldehyde Azine, Mikromol™
Artikelnummer. 30051780
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051780 100 mg
30051204 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051780 Leverandør Mikromol Leverandørnr. MM0905.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-(4-Nitrophenyl)-2-furaldehyde Azine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(4-nitrophenyl)-2-furaldehyde azine
CAS 301359-05-7
Analyt- eller komponentnavne 5-(4-nitrophenyl)-2-furaldehyde azine
Forsendelsestilstand Room Temperature
Molekylær formel C22 H14 N4 O6
Certificeringer/compliance ISO 9001
Synonym 5-(4-nitrophenyl)-2-furaldehyde azine, 2-Furancarboxaldehyde, 5-(4-nitrophenyl)-, 2-[[5-(4-nitrophenyl)-2-furanyl]methylene]hydrazone, 2-Furancarboxaldehyde, 5-(4-nitrophenyl)-, [[5-(4-nitrophenyl)-2-furanyl]methylene]hydrazone (9CI), Dantrolene Related Compound A
InChI formel InChI=1S/C22H14N4O6/c27-25(28)17-5-1-15(2-6-17)21-11-9-19(31-21)13-23-24-14-20-10-12-22(32-20)16-3-7-18(8-4-16)26(29)30/h1-14H
SMIL [O-][N+](=O)c1ccc(cc1)c2oc(C=NN=Cc3oc(cc3)c4ccc(cc4)[N+](=O)[O-])cc2
IUPAC navn 1-[5-(4-nitrophenyl)furan-2-yl]-N-[[5-(4-nitrophenyl)furan-2-yl]methylideneamino]methanimine
Molekylvægt (g/mol) 430.37
Mængde 100 mg
Formel vægt 430.0913 g/mol