Få mere at vide
5-(4-Morpholinylmethyl)-d2 Delta-Tocopherol, 90%, TRC
Artikelnummer. 30471264
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30471264 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471264 Leverandør Toronto Research Chemicals Leverandørnr. TRCM7237925MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(4-Morpholinylmethyl)-d2 delta-Tocopherol
CAS 936230-74-9
Molekylær formel C32H53D2NO3
Mængde 5 mg
Synonym (2R)-3,4-Dihydro-2,8-dimethyl-5-(4-morpholinylmethyl-d2)-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-ol
InChI formel InChI=1S/C32H55NO3/c1-24(2)10-7-11-25(3)12-8-13-26(4)14-9-16-32(6)17-15-28-29(23-33-18-20-35-21-19-33)30(34)22-27(5)31(28)36-32/h22,24-26,34H,7-21,23H2,1-6H3/t25-,26-,32-/m1/s1/i23D2
SMIL OC1=C(C([2H])([2H])N2CCOCC2)C(CC[C@](CCCC(C)CCCC(C)CCCC(C)C)(C)O3)=C3C(C)=C1
Molekylvægt (g/mol) 503.8