Få mere at vide
5-(4-Methoxy-d3-2,3,6-trimethylphenyl)-3-methyl-2,4-pentadien-1-ol, TRC
Artikelnummer. 30473821
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30473821 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473821 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2716225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(4-Methoxy-d3-2,3,6-trimethylphenyl)-3-methyl-2,4-pentadien-1-ol
CAS 1185234-83-6
Molekylær formel C16 D3 H19 O2
Mængde 5 mg
InChI formel InChI=1S/C16H22O2/c1-11(8-9-17)6-7-15-12(2)10-16(18-5)14(4)13(15)3/h6-8,10,17H,9H2,1-5H3/b7-6+,11-8+/i5D3
SMIL [2H]C([2H])([2H])Oc1cc(C)c(\C=C\C(=C\CO)\C)c(C)c1C
IUPAC navn (2E,4E)-3-methyl-5-[2,3,6-trimethyl-4-(trideuteriomethoxy)phenyl]penta-2,4-dien-1-ol
Molekylvægt (g/mol) 249.363
Formel vægt 249.181