Få mere at vide
5-(4-Hydroxyphenyl)-5-phenylhydantoin, TRC
Artikelnummer. 30395276
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30395276 100 mg
30392212 25 mg
30395662 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30395276 Leverandør Toronto Research Chemicals Leverandørnr. TRCH516770100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2784-27-2
Molekylær formel C15 H12 N2 O3
Molekylvægt (g/mol) 268.2674
Synonym Hydantoin, 5-(p-hydroxyphenyl)-5-phenyl-, (p-Hydroxyphenyl)phenylhydantoin, (±)-5-(4-Hydroxyphenyl-5-phenylhydantoin, 4-Hydroxydiphenylhydantoin, 4-Hydroxyphenytoin, 5-(4-Hydroxyphenyl)-5-phenylhydantoin, 5-(4'-Hydroxyphenyl)-5-phenylhydantoin, 5-(p-Hydroxyphenyl)-5-phenylhydantoin, 5-Phenyl-5-p-hydroxyphenylhydantoin, Hydroxydiphenylhydantoin, NSC 156081, dl-5-(p-Hydroxyphenyl-5-phenylhydantoin, p-Hydroxydiphenylhydantoin, p-Hydroxyphenytoin, 2,4-Imidazolidinedione, 5-(4-hydroxyphenyl)-5-phenyl-
IUPAC navn 5-(4-hydroxyphenyl)-5-phenylimidazolidine-2,4-dione
SMIL Oc1ccc(cc1)C2(NC(=O)NC2=O)c3ccccc3
Tekniske data
Kemisk navn eller materiale 5-(4-Hydroxyphenyl)-5-phenylhydantoin
Mængde 100 mg
InChI formel InChI=1S/C15H12N2O3/c18-12-8-6-11(7-9-12)15(10-4-2-1-3-5-10)13(19)16-14(20)17-15/h1-9,18H,(H2,16,17,19,20)
Formel vægt 268.0848