missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-Fluorophenyl)cyclohexane-1,3-dione, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 206.21 |
Percent Purity | 95% |
Chemical Name or Material | 5-(4-Fluorophenyl)cyclohexane-1, 3-dione |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'