Få mere at vide
5-(4'-(Dibromomethyl)-[1,1'-biphenyl]-2-yl)-1H-tetrazole, TRC
Artikelnummer. 17977824
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17977824 50 mg
17937534 100 mg
17929164 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17977824 Leverandør Toronto Research Chemicals Leverandørnr. TRCD25576050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1127249-17-5
Molekylær formel C14 H10 Br2 N4
Molekylvægt (g/mol) 394.06
Synonym Olmesartan Impurity 29
IUPAC navn 5-[2-[4-(dibromomethyl)phenyl]phenyl]-1H-tetrazole
SMIL BrC(Br)c1ccc(cc1)c2ccccc2c3nnn[nH]3
Tekniske data
Kemisk navn eller materiale 5-(4'-(Dibromomethyl)-[1,1'-biphenyl]-2-yl)-1H-tetrazole
Mængde 50 mg
InChI formel InChI=1S/C14H10Br2N4/c15-13(16)10-7-5-9(6-8-10)11-3-1-2-4-12(11)14-17-19-20-18-14/h1-8,13H,(H,17,18,19,20)
Formel vægt 391.9272