Få mere at vide
5-(4-Benzyloxyphenyl)nicotinic acid, TRC
Artikelnummer. 30340118
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30340118 250 mg
30330507 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30340118 Leverandør Toronto Research Chemicals Leverandørnr. TRCB288923250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(4-Benzyloxyphenyl)nicotinic acid
CAS 893739-18-9
Molekylær formel C19 H15 N O3
Mængde 250 mg
InChI formel InChI=1S/C19H15NO3/c21-19(22)17-10-16(11-20-12-17)15-6-8-18(9-7-15)23-13-14-4-2-1-3-5-14/h1-12H,13H2,(H,21,22)
SMIL OC(=O)c1cncc(c1)c2ccc(OCc3ccccc3)cc2
IUPAC navn 5-(4-phenylmethoxyphenyl)pyridine-3-carboxylic acid
Molekylvægt (g/mol) 305.327
Formel vægt 305.105