missing translation for 'onlineSavingsMsg'
Få mere at vide

5-(4’-Benzyloxyphenyl)-2-oxazolidone, TRC

Product Code. 30342300
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30342300 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342300 Supplier Toronto Research Chemicals Supplier No. TRCB28771525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-(4'-Benzyloxyphenyl)-2-oxazolidone
CAS 88693-98-5
Molekylær formel C16 H15 N O3
Mængde 25 mg
InChI formel InChI=1S/C16H15NO3/c18-16-17-10-15(20-16)13-6-8-14(9-7-13)19-11-12-4-2-1-3-5-12/h1-9,15H,10-11H2,(H,17,18)
SMIL O=C1NCC(O1)c2ccc(OCc3ccccc3)cc2
IUPAC navn 5-(4-phenylmethoxyphenyl)-1,3-oxazolidin-2-one
Molekylvægt (g/mol) 269.3
Formel vægt 269.1052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.