Få mere at vide
5-[[4-[3-(Aminocarbonyl)-1,4,5,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo-6H-pyrazolo[3,4-c]pyridin-6-yl]phenyl]amino]pentanoic Acid, TRC
Artikelnummer. 30132455
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132455 100 mg
17914885 10 mg
17980385 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132455 Leverandør Toronto Research Chemicals Leverandørnr. TRCA875620100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2206825-87-6
Molekylær formel C25 H27 N5 O5
Molekylvægt (g/mol) 477.51
Synonym Pentanoic acid, 5-[[4-[3-(aminocarbonyl)-1,4,5,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo-6H-pyrazolo[3,4-c]pyridin-6-yl]phenyl]amino]-, 5-[[4-[3-(Aminocarbonyl)-1,4,5,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo-6H-pyrazolo[3,4-c]pyridin-6-yl]phenyl]amino]pentanoic acid
IUPAC navn 5-[4-[3-carbamoyl-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridin-6-yl]anilino]pentanoic acid
SMIL COc1ccc(cc1)n2nc(C(=O)N)c3CCN(C(=O)c23)c4ccc(NCCCCC(=O)O)cc4
Tekniske data
Kemisk navn eller materiale 5-((4-(3-(Aminocarbonyl)-1,4,5,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo-6H-pyrazolo(3,4-c)pyridin-6-yl)phenyl)amino)pentanoic Acid
Analyt- eller komponentnavne 5-((4-(3-(Aminocarbonyl)-1,4,5,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo-6H-pyrazolo(3,4-c)pyridin-6-yl)phenyl)amino)pentanoic Acid
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C25H27N5O5/c1-35-19-11-9-18(10-12-19)30-23-20(22(28-30)24(26)33)13-15-29(25(23)34)17-7-5-16(6-8-17)27-14-3-2-4-21(31)32/h5-12,27H,2-4,13-15H2,1H3,(H2,26,33)(H,31,32)
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 100 mg
Formel vægt 477.2012
Procent renhed >95%