Få mere at vide
5-[4-[2-[Methyl(pyridin-2-yl)amino]ethoxy]benzyl]-2-imino-1,3-thiazolidin-4-one, Mikromol™
Artikelnummer. 30052694
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052694 25 mg
30050776 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052694 Leverandør Mikromol Leverandørnr. MM0522.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-[4-[2-[Methyl(pyridin-2-yl)amino]ethoxy]benzyl]-2-imino-1,3-thiazolidin-4-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-[4-[2-[Methyl(pyridin-2-yl)amino]ethoxy]benzyl]-2-imino-1,3-thiazolidin-4-one
CAS 329249-53-8
Analyt- eller komponentnavne 5-[4-[2-[Methyl(pyridin-2-yl)amino]ethoxy]benzyl]-2-imino-1,3-thiazolidin-4-one
Forsendelsestilstand Room Temperature
Molekylær formel C18 H20 N4 O2 S
Certificeringer/compliance ISO 9001
Synonym 5-[4-[2-[Methyl(pyridin-2-yl)amino]ethoxy]benzyl]-2-imino-1,3-thiazolidin-4-one
InChI formel InChI=1S/C18H20N4O2S/c1-22(16-4-2-3-9-20-16)10-11-24-14-7-5-13(6-8-14)12-15-17(23)21-18(19)25-15/h2-9,15H,10-12H2,1H3,(H2,19,21,23)
SMIL CN(CCOc1ccc(CC2SC(=N)NC2=O)cc1)c3ccccn3
IUPAC navn 2-imino-5-[[4-[2-[methyl(pyridin-2-yl)amino]ethoxy]phenyl]methyl]-1,3-thiazolidin-4-one
Molekylvægt (g/mol) 356.44
Mængde 25 mg
Formel vægt 356.1307 g/mol