Få mere at vide
5-[4-[2-((Methyl-d3)pyridin-2-ylamino)ethoxy]benzylidene]thiazolidine-2,4-dione, TRC
Artikelnummer. 30466490
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30466490 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466490 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32600725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-[[4-[2-((Methyl-d3)-2-pyridinylamino)ethoxy]phenyl]methylene]-2,4-thiazolidinedione
CAS 1185245-57-1
Molekylær formel C18H14D3N3O3S
Mængde 25 mg
InChI formel InChI=1S/C18H17N3O3S/c1-21(16-4-2-3-9-19-16)10-11-24-14-7-5-13(6-8-14)12-15-17(22)20-18(23)25-15/h2-9,12H,10-11H2,1H3,(H,20,22,23)/b15-12+/i1D3
SMIL [2H]C([2H])([2H])N(CCOc1ccc(\C=C/2\SC(=O)NC2=O)cc1)c3ccccn3
IUPAC navn (5E)-5-[[4-[2-[pyridin-2-yl(trideuteriomethyl)amino]ethoxy]phenyl]methylidene]-1,3-thiazolidine-2,4-dione
Molekylvægt (g/mol) 358.43
Formel vægt 358.1179