Få mere at vide
5-[[4-[2-(5-Ethyl-2-pyridinyl)ethoxy]phenyl]-methylene]-2,4-thiazolidinedione(Pioglitazone Impurity), TRC
Artikelnummer. 30136283
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136283 25 mg
30136139 50 mg
30010971 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136283 Leverandør Toronto Research Chemicals Leverandørnr. TRCE92559025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 144809-28-9
Molekylær formel C19 H18 N2 O3 S
Molekylvægt (g/mol) 354.42
Synonym 2,4-Thiazolidinedione, 5-[[4-[2-(5-ethyl-2-pyridinyl)ethoxy]phenyl]methylene]-, 5-[[4-[2-(5-Ethyl-2-pyridyl)ethoxy]phenyl]methylene]thiazolidine-2,4-dione
IUPAC navn (5E)-5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methylidene]-1,3-thiazolidine-2,4-dione
SMIL CCc1ccc(CCOc2ccc(\C=C/3\SC(=O)NC3=O)cc2)nc1
Tekniske data
Kemisk navn eller materiale 5-[(4-[2-(5-Ethyl-2-pyridinyl)ethoxy]phenyl)methylene]-2,4-thiazolidinedione
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C19H18N2O3S/c1-2-13-3-6-15(20-12-13)9-10-24-16-7-4-14(5-8-16)11-17-18(22)21-19(23)25-17/h3-8,11-12H,2,9-10H2,1H3,(H,21,22,23)/b17-11+
Formel vægt 354.1038
Procent renhed >95%