missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Pyridyl)-1H-tetrazole, 98%, Thermo Scientific Chemicals

Catalog Number p-7025145
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3250-74-6
C6H5N5
147.141
MFCD00040281
SECHDFHDDVELCV-UHFFFAOYSA-N
3-2h-tetrazol-5-yl pyridine, 5-3-pyridyl-1h-tetrazole, 3-1h-tetrazol-5-yl pyridine, 5-3-pyridyl tetrazole, 3-2h-tetrazol-5-yl-pyridine, pyridine, 3-2h-tetrazol-5-yl, unii-zc84pps834, 3-2h-1,2,3,4-tetrazol-5-yl pyridine, 3-1h-1,2,3,4-tetrazol-5-yl pyridine, 5-3-pyridyl-2h-1,2,3,4-tetraazole
151091
3-(2H-tetrazol-5-yl)pyridine
C1=CC(=CN=C1)C2=NNN=N2
This item is not returnable. View return policy
3250-74-6
C6H5N5
147.141
MFCD00040281
SECHDFHDDVELCV-UHFFFAOYSA-N
3-2h-tetrazol-5-yl pyridine, 5-3-pyridyl-1h-tetrazole, 3-1h-tetrazol-5-yl pyridine, 5-3-pyridyl tetrazole, 3-2h-tetrazol-5-yl-pyridine, pyridine, 3-2h-tetrazol-5-yl, unii-zc84pps834, 3-2h-1,2,3,4-tetrazol-5-yl pyridine, 3-1h-1,2,3,4-tetrazol-5-yl pyridine, 5-3-pyridyl-2h-1,2,3,4-tetraazole
151091
3-(2H-tetrazol-5-yl)pyridine
C1=CC(=CN=C1)C2=NNN=N2

5-(3-Pyridyl)-1H-tetrazole is used as a pharmaceutical intermediate. The Pyridine,3-(2H-tetrazol-5-yl)- acts as reactant with 1-(4-bromo-phenyl)-3-piperidino-propan-1-one, hydrochloride, and obtain the 1-(4-bromo-phenyl)-3-(5-pyridin-3-yl-tetrazol-2-yl)-propan-1-one. This reaction needs the solvent of dimethylformamide. The yield is 31%. In addition, this reaction should be taken for 8 hours at the temperature of 150°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(3-Pyridyl)-1H-tetrazole is used as a pharmaceutical intermediate. The Pyridine,3-(2H-tetrazol-5-yl)- acts as reactant with 1-(4-bromo-phenyl)-3-piperidino-propan-1-one, hydrochloride, and obtain the 1-(4-bromo-phenyl)-3-(5-pyridin-3-yl-tetrazol-2-yl)-propan-1-one. This reaction needs the solvent of dimethylformamide. The yield is 31%. In addition, this reaction should be taken for 8 hours at the temperature of 150°C.

Solubility
Soluble in water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼240°C (decomposition)
Odor Odorless
Quantity 1 g
Solubility Information Soluble in water (slightly).
Formula Weight 147.14
Percent Purity 98%
Chemical Name or Material 5-(3-Pyridyl)-1H-tetrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'