missing translation for 'onlineSavingsMsg'
Få mere at vide

5-(3-Bromopropyl)-5H-dibenz[b,f]azepine, TRC

Artikelnummer. 30330300
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30330300 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30330300 Leverandør Toronto Research Chemicals Leverandørnr. TRCB282230500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-(3-Bromopropyl)-5H-dibenz[b,f]azepine
CAS 62035-53-4
Molekylær formel C17 H16 Br N
Mængde 500 mg
Synonym 5-(3-Bromopropyl)-5H-dibenz[b,f]azepine, n-(Bromopropyl)iminostilbene
InChI formel InChI=1S/C17H16BrN/c18-12-5-13-19-16-8-3-1-6-14(16)10-11-15-7-2-4-9-17(15)19/h1-4,6-11H,5,12-13H2
SMIL BrCCCN1c2ccccc2C=Cc3ccccc13
IUPAC navn 11-(3-bromopropyl)benzo[b][1]benzazepine
Molekylvægt (g/mol) 314.22
Formel vægt 313.047
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.