Få mere at vide
5-(3-Bromo-4,5-dimethoxybenzyl)pyrimidine-2,4-diamine, Mikromol™
Artikelnummer. 30049452
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049452 25 mg
17941686 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049452 Leverandør Mikromol Leverandørnr. MM0093.090025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-(3-Bromo-4,5-dimethoxybenzyl)pyrimidine-2,4-diamine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(3-Bromo-4,5-dimethoxybenzyl)pyrimidine-2,4-diamine
CAS 16285-82-8
Analyt- eller komponentnavne 5-(3-Bromo-4,5-dimethoxybenzyl)pyrimidine-2,4-diamine
Forsendelsestilstand Room Temperature
Molekylær formel C13 H15 Br N4 O2
Certificeringer/compliance ISO 17025
Synonym Trimethoprim Imp. F (EP), 5-(3-Bromo-4,5-dimethoxybenzyl)pyrimidine-2,4-diamine, Trimethoprim Impurity F
InChI formel InChI=1S/C13H15BrN4O2/c1-19-10-5-7(4-9(14)11(10)20-2)3-8-6-17-13(16)18-12(8)15/h4-6H,3H2,1-2H3,(H4,15,16,17,18)
SMIL COc1cc(Cc2cnc(N)nc2N)cc(Br)c1OC
IUPAC navn 5-[(3-bromo-4,5-dimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Molekylvægt (g/mol) 339.19
Mængde 25 mg
Formel vægt 338.0378 g/mol