missing translation for 'onlineSavingsMsg'
Learn More

5-(3,4-Dichlorophenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15490826
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1449142-43-1
C13H9BCl2FNO3
327.927
MFCD20040143
QBXYLSUKPFQIGC-UHFFFAOYSA-N
5-3,4-dichlorophenylcarbamoyl-2-fluorobenzeneboronic acid, 5-3,4-dichlorophenyl carbamoyl-2-fluorophenylboronic acid
73995661
[5-[(3,4-dichlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CC(=C(C=C2)Cl)Cl)F)(O)O
This item is not returnable. View return policy
1449142-43-1
C13H9BCl2FNO3
327.927
MFCD20040143
QBXYLSUKPFQIGC-UHFFFAOYSA-N
5-3,4-dichlorophenylcarbamoyl-2-fluorobenzeneboronic acid, 5-3,4-dichlorophenyl carbamoyl-2-fluorophenylboronic acid
73995661
[5-[(3,4-dichlorophenyl)carbamoyl]-2-fluorophenyl]boronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=CC(=C(C=C2)Cl)Cl)F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 327.93
Percent Purity 97%
Chemical Name or Material 5-(3,4-Dichlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'