Få mere at vide
5-[3-(2-Acetyl-3-hydroxyphenoxy)-2-hydroxypropoxy]-4-oxo-4H-1-benzopyran-2-carboxylic Acid Sodium Salt, Mikromol™
Artikelnummer. 30051831
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051831 25 mg
17960426 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051831 Leverandør Mikromol Leverandørnr. MM0039.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-[3-(2-Acetyl-3-hydroxyphenoxy)-2-hydroxypropoxy]-4-oxo-4H-1-benzopyran-2-carboxylic Acid Sodium Salt
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-[3-(2-Acetyl-3-hydroxyphenoxy)-2-hydroxypropoxy]-4-oxo-4H-1-benzopyran-2-carboxylic Acid Sodium Salt
CAS 40820-38-0
Analyt- eller komponentnavne 5-[3-(2-Acetyl-3-hydroxyphenoxy)-2-hydroxypropoxy]-4-oxo-4H-1-benzopyran-2-carboxylic Acid Sodium Salt
Forsendelsestilstand Room Temperature
Molekylær formel C21 H17 O9 . Na
Certificeringer/compliance ISO 17025
Synonym 5-[3-(2-Acetyl-3-hydroxyphenoxy)-2-hydroxypropoxy]-4-oxo-4H-1-benzopyran-2-carboxylic Acid Sodium Salt (1:1), 5-[3-(2-Acetyl-3-hydroxyphenoxy)-2-hydroxypropoxy]-4-oxo-4H-1-benzopyran-2-carboxylic Acid Monosodium Salt (9CI)
InChI formel InChI=1S/C21H18O9.Na/c1-11(22)19-13(24)4-2-5-15(19)28-9-12(23)10-29-16-6-3-7-17-20(16)14(25)8-18(30-17)21(26)27;/h2-8,12,23-24H,9-10H2,1H3,(H,26,27);/q;+1/p-1
SMIL [Na+].CC(=O)c1c(O)cccc1OCC(O)COc2cccc3OC(=CC(=O)c23)C(=O)[O-]
IUPAC navn sodium;5-[3-(2-acetyl-3-hydroxyphenoxy)-2-hydroxypropoxy]-4-oxochromene-2-carboxylate
Molekylvægt (g/mol) 436.34
Mængde 25 mg
Formel vægt 436.077 g/mol