Få mere at vide
5-[(2R)-2-[[2-(2-Hydroxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride (Desethyltamsulosin Hydrochloride), Mikromol™
Artikelnummer. 30051642
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051642 25 mg
17923796 100 mg
30462023 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30051642 Leverandør Mikromol Leverandørnr. MM0938.160025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-[(2R)-2-[[2-(2-Hydroxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride (Desethyltamsulosin Hydrochloride)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desethyltamsulosin Hydrochloride
CAS 1582717-04-1
Analyt- eller komponentnavne Desethyltamsulosin Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C18 H24 N2 O5 S . Cl H
Certificeringer/compliance ISO 17025
Synonym 5-[(2R)-2-[[2-(2-Hydroxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride (1:1), Desethyltamsulosin Hydrochloride
InChI formel InChI=1S/C18H24N2O5S.ClH/c1-13(20-9-10-25-16-6-4-3-5-15(16)21)11-14-7-8-17(24-2)18(12-14)26(19,22)23;/h3-8,12-13,20-21H,9-11H2,1-2H3,(H2,19,22,23);1H/t13-;/m1./s1
SMIL Cl.COc1ccc(C[C@@H](C)NCCOc2ccccc2O)cc1S(=O)(=O)N
IUPAC navn 5-[(2R)-2-[2-(2-hydroxyphenoxy)ethylamino]propyl]-2-methoxybenzenesulfonamide;hydrochloride
Molekylvægt (g/mol) 416.92
Mængde 25 mg
Formel vægt 416.1173 g/mol