Få mere at vide
5-(2-Nitro-1-propenyl)-1,3-benzodioxole, TRC
Artikelnummer. 30483816
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483816 100 mg
30465142 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30483816 Leverandør Toronto Research Chemicals Leverandørnr. TRCN519705100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(3,4-Methylenedioxyphenyl)-2-nitropropene
CAS 5438-41-5
Molekylær formel C10 H9 N O4
Mængde 100 mg
Synonym 5-(2-Nitro-1-propen-1-yl)-1,3-Benzodioxole, 1,3-Benzodioxole','5-(2-nitro-1-propenyl)- (9CI), Benzene','1,2-(methylenedioxy)-4-(2-nitropropenyl)- (6CI,7CI,8CI), 5-(2-Nitro-1-propen-1-yl)-1,3-benzodioxole, 1,2-Methylenedioxy-4-(2-nitropropenyl)benzene, 1-(2-Nitro-1-propenyl)-3,4-methylenedioxybenzene, 2-Nitro-1-(3,4-methylenedioxyphenyl)-1-propene, 3,4-Methylenedioxy-1-(2-nitropropenyl)benzene, Iksin, NSC 16685, NSC 65667, β-Methyl-β-nitro-3,4-methylenedioxystyrene
InChI formel InChI=1S/C10H9NO4/c1-7(11(12)13)4-8-2-3-9-10(5-8)15-6-14-9/h2-5H,6H2,1H3
SMIL CC(=Cc1ccc2OCOc2c1)[N+](=O)[O-]
IUPAC navn 5-(2-nitroprop-1-enyl)-1,3-benzodioxole
Molekylvægt (g/mol) 207.1
Formel vægt 207.1828