Få mere at vide
5-[2-Ethoxy-5-[(4-methyl-1-piperazinyl)thioxomethyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidine-7-thione, TRC
Artikelnummer. 30372015
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30372015 10 mg
30370454 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30372015 Leverandør Toronto Research Chemicals Leverandørnr. TRCE89253010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1333233-46-7
Molekylær formel C23 H30 N6 O S2
Molekylvægt (g/mol) 470.65
Synonym 7H-Pyrazolo[4,3-d]pyrimidine-7-thione, 5-[2-ethoxy-5-[(4-methyl-1-piperazinyl)thioxomethyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-
IUPAC navn 5-[2-ethoxy-5-(4-methylpiperazine-1-carbothioyl)phenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidine-7-thione
SMIL CCCc1nn(C)c2C(=S)NC(=Nc12)c3cc(ccc3OCC)C(=S)N4CCN(C)CC4
Tekniske data
Kemisk navn eller materiale Dithio-desmethyl-carbodenafil
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C23H30N6OS2/c1-5-7-17-19-20(28(4)26-17)22(31)25-21(24-19)16-14-15(8-9-18(16)30-6-2)23(32)29-12-10-27(3)11-13-29/h8-9,14H,5-7,10-13H2,1-4H3,(H,24,25,31)
Formel vægt 470.1923
Procent renhed >95%