missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Chlorophenyl)-1H-tetrazole, 98%, Thermo Scientific™

Catalog Number 11387596
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
50907-46-5
C7H5ClN4
180.60
MFCD00040950
PSUIIKIEUATWCZ-UHFFFAOYSA-N
5-2-chlorophenyl-1h-tetrazole, 5-2-chlorophenyl-2h-tetrazole, 5-2-chlorophenyl-2h-1,2,3,4-tetrazole, 5-2-chlorophenyl tetrazole, 5-2-chlorophenyl-2h-tetraazole, 2h-tetrazole,5-2-chlorophenyl, 5-2-chlorophenyl-1h-1,2,3,4-tetrazole, 5-2-chlorophenyl-1h-1,2,3,4-tetraazole, ldha inhibitor, 14, 2-chlorophenyl tetrazole
560735
ClC1=CC=CC=C1C1=NNN=N1
This item is not returnable. View return policy
50907-46-5
C7H5ClN4
180.60
MFCD00040950
PSUIIKIEUATWCZ-UHFFFAOYSA-N
5-2-chlorophenyl-1h-tetrazole, 5-2-chlorophenyl-2h-tetrazole, 5-2-chlorophenyl-2h-1,2,3,4-tetrazole, 5-2-chlorophenyl tetrazole, 5-2-chlorophenyl-2h-tetraazole, 2h-tetrazole,5-2-chlorophenyl, 5-2-chlorophenyl-1h-1,2,3,4-tetrazole, 5-2-chlorophenyl-1h-1,2,3,4-tetraazole, ldha inhibitor, 14, 2-chlorophenyl tetrazole
560735
ClC1=CC=CC=C1C1=NNN=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 179°C to 180°C
Beilstein 148559
IUPAC Name 5-(2-chlorophenyl)-2H-1,2,3,4-tetrazole
Formula Weight 180.6
Percent Purity 98%
Quantity 1 g
Chemical Name or Material 5-(2-Chlorophenyl)-1H-tetrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'