Få mere at vide
5-[(2-Chloroethyl)(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester, TRC
Artikelnummer. 30366558
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366558 100 mg
30358154 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30366558 Leverandør Toronto Research Chemicals Leverandørnr. TRCC369650100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 898224-95-8
Molekylær formel C18H26ClN3O3
Molekylvægt (g/mol) 367.87
Synonym Bendamustine Impurity
IUPAC navn ethyl 4-(5-((2-chloroethyl)(2-hydroxyethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoate
SMIL OCCN(CCCl)C1=CC(N=C(CCCC(OCC)=O)N2C)=C2C=C1
Tekniske data
Kemisk navn eller materiale 5-[(2-Chloroethyl)(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Ethyl Ester
Mængde 100 mg
InChI formel InChI=1S/C18H26ClN3O3/c1-3-25-18(24)6-4-5-17-20-15-13-14(7-8-16(15)21(17)2)22(10-9-19)11-12-23/h7-8,13,23H,3-6,9-12H2,1-2H3
Formel vægt 367.17