Få mere at vide
5-(2-Chlorobenzyl)thieno[3,2-c]pyridinium Chloride, Mikromol™
Beskrivelse
5-(2-Chlorobenzyl)thieno[3,2-c]pyridinium Chloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 5-(2-Chlorobenzyl)thieno[3,2-c]pyridinium Chloride |
| CAS | 53885-64-6 |
| Analyt- eller komponentnavne | 5-(2-Chlorobenzyl)thieno[3,2-c]pyridinium Chloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H11 Cl N S . Cl |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Ticlopidine Hydrochloride Imp. E (EP), Ticlopidine Imp. E (EP), 5-(2-Chlorobenzyl)thieno[3,2-c]pyridinium Chloride, Ticlopidine Hydrochloride Imp. E (EP) as Chloride, Ticlopidine Hydrochloride Impurity E as Chloride, Ticlopidine Impurity E as Chloride |
| InChI formel | InChI=1S/C14H11ClNS.ClH/c15-13-4-2-1-3-11(13)9-16-7-5-14-12(10-16)6-8-17-14;/h1-8,10H,9H2;1H/q+1;/p-1 |
| SMIL | [Cl-].Clc1ccccc1C[n+]2ccc3sccc3c2 |
| IUPAC navn | 5-[(2-chlorophenyl)methyl]thieno[3,2-c]pyridin-5-ium;chloride |
| Vis mere |