missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-(2-Bromophenyl)-1H-tetrazole, 98+%
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Melting Point | 181°C to 183°C |
Quantity | 1 g |
Beilstein | 148567 |
Synonym | 5-2-bromophenyl-1h-tetrazole, 5-2-bromophenyl tetrazole, 5-2-bromo-phenyl-2h-tetrazole, 5-2-bromophenyl-2h-tetrazole, 5-2-bromophenyl-1h-1,2,3,4-tetrazole, 5-2-bromophenyl-2h-1,2,3,4-tetrazole, 5-2-bromophenyl-1h-tetraazole, 5-2-bromophenyl-1 h-tetrazole, 1h-tetrazole, 5-2-bromophenyl |
Formula Weight | 225.05 |
Percent Purity | ≥98% |
Chemical Name or Material | 5-(2-Bromophenyl)-1H-tetrazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'