Få mere at vide
5-(2-Bromoethyl-d4)-2,3-dihydrobenzofuran, TRC
Artikelnummer. 30343235
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30343235 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343235 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68423410MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(2-Bromoethyl-d4)-2,3-dihydrobenzofuran
CAS 1189652-25-2
Molekylær formel C10H7D4BrO
Mængde 10 mg
InChI formel InChI=1S/C10H11BrO/c11-5-3-8-1-2-10-9(7-8)4-6-12-10/h1-2,7H,3-6H2/i3D2,5D2
SMIL [2H]C([2H])(Br)C([2H])([2H])c1ccc2OCCc2c1
IUPAC navn 5-(2-bromo-1,1,2,2-tetradeuterioethyl)-2,3-dihydro-1-benzofuran
Molekylvægt (g/mol) 231.12
Formel vægt 230.0244