missing translation for 'onlineSavingsMsg'
Få mere at vide

5-(2-Bromoethyl)-2,3-dihydrobenzofuran, 98%, BLD Pharm™

Artikelnummer. 31089926
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 g
5 g
10 g
25 g
100 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31089926 10 g
31079014 1 g
31085010 100 g
31079594 250 mg
31078749 25 g
31088228 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31089926 Leverandør BLD Pharm Leverandørnr. BD1186710g
 Begrænset vare
Forventet forsendelse: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale 5-(2-Bromoethyl)-2,3-dihydrobenzofuran
CAS 127264-14-6
Molekylær formel C10H11BrO
MDL nummer MFCD06797641
Mængde 10 g
SMIL BrCCC1=CC=C(OCC2)C2=C1
Molekylvægt (g/mol) 227.10
Procent renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.