missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-(2,5-Dichlorophenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%
Brand: Alfa Aesar™ H54460.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
2096335-17-8 | |
327.93 | |
NRCZOQMPVBOVGS-UHFFFAOYSA-N | |
73995818 |
C13H9BCl2FNO3 | |
MFCD20040191 | |
5-2,5-dichlorophenylcarbamoyl-2-fluorobenzeneboronic acid |
Specifications
5-(2,5-Dichlorophenylcarbamoyl)-2-fluorobenzeneboronic acid | |
2096335-17-8 | |
MFCD20040191 | |
NRCZOQMPVBOVGS-UHFFFAOYSA-N | |
73995818 | |
97% |
1g | |
C13H9BCl2FNO3 | |
5-2,5-dichlorophenylcarbamoyl-2-fluorobenzeneboronic acid | |
327.93 | |
327.93 |