Få mere at vide
5-[2-(2-Naphthyloxy)phenyl]-2H-tetrazole, TRC
Artikelnummer. 30421955
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421955 10 mg
30415564 100 mg
30412000 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30421955 Leverandør Toronto Research Chemicals Leverandørnr. TRCN92626510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1305320-61-9
Molekylær formel C17H12N4O
Molekylvægt (g/mol) 288.3
IUPAC navn 5-(2-naphthalen-2-yloxyphenyl)-2H-tetrazole
SMIL O(c1ccc2ccccc2c1)c3ccccc3c4nn[nH]n4
Tekniske data
Kemisk navn eller materiale 5-[2-(2-Naphthyloxy)phenyl]-2H-tetrazole
Mængde 10 mg
InChI formel InChI=1S/C17H12N4O/c1-2-6-13-11-14(10-9-12(13)5-1)22-16-8-4-3-7-15(16)17-18-20-21-19-17/h1-11H,(H,18,19,20,21)
Formel vægt 288.1011