missing translation for 'onlineSavingsMsg'
Få mere at vide

5-[(1RS)-1-Hydroxyethyl]-1,3-phenylene Bis(dimethylcarbamate), Mikromol™

Artikelnummer. 30052067
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052067 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30052067 Leverandør Mikromol Leverandørnr. MM0773.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-[(1RS)-1-Hydroxyethyl]-1,3-phenylene Bis(dimethylcarbamate)
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-[(1RS)-1-Hydroxyethyl]-1,3-phenylene Bis(dimethylcarbamate)
CAS 112935-93-0
Analyt- eller komponentnavne 5-[(1RS)-1-Hydroxyethyl]-1,3-phenylene Bis(dimethylcarbamate)
Forsendelsestilstand Room Temperature
Molekylær formel C14 H20 N2 O5
Certificeringer/compliance ISO/IEC 17025
Synonym Bambuterol Hydrochloride Imp. D (EP), Bambuterol Imp. D (EP), 5-[(1RS)-1-Hydroxyethyl]-1,3-phenylene Bis(dimethylcarbamate), Bambuterol Hydrochloride Impurity D, Bambuterol Impurity D
InChI formel InChI=1S/C14H20N2O5/c1-9(17)10-6-11(20-13(18)15(2)3)8-12(7-10)21-14(19)16(4)5/h6-9,17H,1-5H3
SMIL CC(O)c1cc(OC(=O)N(C)C)cc(OC(=O)N(C)C)c1
IUPAC navn [3-(dimethylcarbamoyloxy)-5-(1-hydroxyethyl)phenyl] N,N-dimethylcarbamate
Molekylvægt (g/mol) 296.32
Mængde 100 mg
Formel vægt 296.1372 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.