Få mere at vide
5,10-Dihydro-2-methyl-4H-thieno[2,3-b][1,5]benzodiazepin-4-one (Olanzapine Impurity), TRC
Artikelnummer. 30007166
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007166 25 mg
30004108 10 mg
30006768 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007166 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44918525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 221176-49-4
Molekylær formel C12 H10 N2 O S
Molekylvægt (g/mol) 230.29
Synonym 2-Methyl-5,10-dihydro-4H-thieno[2,3-b][1,5]benzodiazepin-4-one, Olanzapine Imp. B (EP), Olanzapine Lactame
IUPAC navn 2-methyl-5,10-dihydrothieno[3,2-c][1,5]benzodiazepin-4-one
SMIL Cc1cc2C(=O)Nc3ccccc3Nc2s1
Tekniske data
Kemisk navn eller materiale Olanzapine Lactame
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C12H10N2OS/c1-7-6-8-11(15)13-9-4-2-3-5-10(9)14-12(8)16-7/h2-6,14H,1H3,(H,13,15)
Formel vægt 230.0514
Procent renhed >95%