Få mere at vide
5,10,15-Tris(4-nitrophenyl)corrole, TRC
Artikelnummer. 30452478
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452478 5 mg
30456395 1 mg
30454386 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30452478 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7951655MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 326472-00-8
Molekylær formel C37H23N7O6
Molekylvægt (g/mol) 661.62
Synonym 1,2,3,7,8,12,13,17,18,19-Decadehydro-21,22-dihydro-5,10,15-tris(4-nitrophenyl)corrin
IUPAC navn (5Z,9Z,14Z)-5,10,15-tris(4-nitrophenyl)-22,24-dihydro-21H-corrin
SMIL [O-][N+](=O)c1ccc(cc1)\C\2=C/3\N\C(=C(\c4ccc(cc4)[N+](=O)[O-])/c5ccc([nH]5)c6ccc([nH]6)\C(=C/7\C=CC2=N7)\c8ccc(cc8)[N+](=O)[O-])\C=C3
Tekniske data
Kemisk navn eller materiale 5,10,15-Tris(4-nitrophenyl)corrole
Mængde 5 mg
InChI formel InChI=1S/C37H23N7O6/c45-42(46)24-7-1-21(2-8-24)35-29-15-13-27(38-29)28-14-16-30(39-28)36(22-3-9-25(10-4-22)43(47)48)32-18-20-34(41-32)37(33-19-17-31(35)40-33)23-5-11-26(12-6-23)44(49)50/h1-20,38-40H/b28-27-,35-29-,35-31-,36-30-,36-32-,37-33-,37-34-
Formel vægt 661.171