Få mere at vide
5-(1-Methylpiperidin-4-yl)-5H-dibenzo[a,d][7]annulen-5-ol Hydrochloride, Mikromol™
Beskrivelse
5-(1-Methylpiperidin-4-yl)-5H-dibenzo[a,d][7]annulen-5-ol Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 5-(1-Methylpiperidin-4-yl)-5H-dibenzo[a,d][7]annulen-5-ol Hydrochloride |
| CAS | 4046-24-6 |
| Analyt- eller komponentnavne | 5-(1-Methylpiperidin-4-yl)-5H-dibenzo[a,d][7]annulen-5-ol Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H23 N O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Cyproheptadine Hydrochloride Imp. C (EP), Cyproheptadine Imp. C (EP), Cyproheptadine USP Related Compound C, Cyproheptadine USP RC C, 5-(1-Methylpiperidin-4-yl)-5H-dibenzo[a,d][7]annulen-5-ol Hydrochloride, Cyproheptadine Hydrochloride Imp. C (EP) as Hydrochloride, Cyproheptadine Related Compound C as Hydrochloride, Cyproheptadine Hydrochloride Impurity C as Hydrochloride, Cyproheptadine Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C21H23NO.ClH/c1-22-14-12-18(13-15-22)21(23)19-8-4-2-6-16(19)10-11-17-7-3-5-9-20(17)21;/h2-11,18,23H,12-15H2,1H3;1H |
| SMIL | Cl.CN1CCC(CC1)C2(O)c3ccccc3C=Cc4ccccc24 |
| IUPAC navn | 11-(1-methylpiperidin-4-yl)dibenzo[1,2-a:2',1'-d][7]annulen-11-ol;hydrochloride |
| Vis mere |