Få mere at vide
(4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazolidine-5-carboxylic Acid, Mikromol™
Artikelnummer. 30049291
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049291 25 mg
30050193 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049291 Leverandør Mikromol Leverandørnr. MM3396.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazolidine-5-carboxylic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazoli
CAS 143527-70-2
Analyt- eller komponentnavne (4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyloxazoli
Forsendelsestilstand Room Temperature
Molekylær formel C17 H23 N O5
Certificeringer/compliance ISO 17025
Synonym 3,5-Oxazolidinedicarboxylic acid, 2,2-dimethyl-4-phenyl-, 3-(1,1-dimethylethyl) ester, (4S,5R)-, 3,5-Oxazolidinedicarboxylic acid, 2,2-dimethyl-4-phenyl-, 3-(1,1-dimethylethyl) ester, (4S-trans)-, (4S,5R)-3-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyl-5-oxazolidinecarboxylic acid, (4S,5R)-3-tert-Butoxycarbonyl-2,2-dimethyl-4-phenyl-1,3-oxazolidine-5-carboxylic acid, (4S,5R)-5-Carboxy-2,2-dimethyl-4-phenyl-3-(tert-butoxycarbonyl)oxazolidine, (4S,5R)-N-(tert-Butoxycarbonyl)-2,2-dimethyl-4-phenyl-5-oxazolidinecarboxylic acid
InChI formel InChI=1S/C17H23NO5/c1-16(2,3)23-15(21)18-12(11-9-7-6-8-10-11)13(14(19)20)22-17(18,4)5/h6-10,12-13H,1-5H3,(H,19,20)/t12-,13+/m0/s1
SMIL CC(C)(C)OC(=O)N1[C@H]([C@@H](OC1(C)C)C(=O)O)c2ccccc2
IUPAC navn (4S,5R)-2,2-dimethyl-3-[(2-methylpropan-2-yl)oxycarbonyl]-4-phenyl-1,3-oxazolidine-5-carboxylic acid
Molekylvægt (g/mol) 321.37
Mængde 25 mg
Formel vægt 321.1576 g/mol