missing translation for 'onlineSavingsMsg'
Få mere at vide

(4S)-4-Methyl-2-oxo-[1,3,2]dioxathiolane-4-carboxylic Acid Methyl Ester, TRC

Artikelnummer. 30464621
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30464621 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30464621 Leverandør Toronto Research Chemicals Leverandørnr. TRCM325845500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (4S)-4-Methyl-2-oxo-[1,3,2]dioxathiolane-4-carboxylic Acid M
CAS 356048-02-7
Molekylær formel C5 H8 O5 S
Mængde 500 mg
InChI formel InChI=1S/C5H8O5S/c1-5(4(6)8-2)3-9-11(7)10-5/h3H2,1-2H3/t5-,11?/m0/s1
SMIL COC(=O)[C@]1(C)COS(=O)O1
IUPAC navn methyl (4S)-4-methyl-2-oxo-1,3,2-dioxathiolane-4-carboxylate
Molekylvægt (g/mol) 180.18
Formel vægt 180.0092
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.