Få mere at vide
(4S)-4-Ethyl-7,8-dihydro-4-hydroxy-1H-pyrano[3,4-f]indolizine-3,6,10(4H)-trione-d5, TRC
Artikelnummer. 30377614
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30377614 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377614 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91730210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4S)-4-Ethyl-7,8-dihydro-4-hydroxy-1H-pyrano[3,4-f]indolizine-3,6,10(4H)-trione-d5
CAS 1346617-23-9
Molekylær formel C13H8D5NO5
Mængde 10 mg
Synonym (S)-4-Ethyl-7,8-dihydro-4-hydroxy-1H-pyrano[3,4-f]indolizine-3,6,10(4H)-trione-d5, (4S)-4-Ethyl-7,8-dihydro-4-hydroxy-1H-pyrano[3,4-f]indolizine-3,6,10(4H)-trione-d5
InChI formel InChI=1S/C13H13NO5/c1-2-13(18)8-5-9-10(15)3-4-14(9)11(16)7(8)6-19-12(13)17/h5,18H,2-4,6H2,1H3/t13-/m0/s1/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])[C@@]1(O)C(=O)OCC2=C1C=C3N(CCC3=O)C2=O
Molekylvægt (g/mol) 268.28