missing translation for 'onlineSavingsMsg'
Learn More

(4S)-4-Cyclohexyl-[(4-phenylbutyl)phosphinyl]acetyl-L-proline, Mikromol™

Product Code. 18083660
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18083660 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18083660 Supplier Mikromol Supplier No. MM0645.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(4S)-4-Cyclohexyl-[(4-phenylbutyl)phosphinyl]acetyl-L-proline
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (4S)-4-Cyclohexyl-[(4-phenylbutyl)phosphinyl]acetyl-L-proline
CAS 95399-71-6
Analyt- eller komponentnavne (4S)-4-Cyclohexyl-[(4-phenylbutyl)phosphinyl]acetyl-L-proline
Forsendelsestilstand Room Temperature
Molekylær formel C23 H34 N O5 P
Certificeringer/compliance ISO 17025
Synonym (4S)-4-Cyclohexyl-1-[[hydroxy(4-phenylbutyl)phosphinyl]acetyl]-L-proline, trans-4-Cyclohexyl-1-[[hydroxy(4-phenylbutyl)phosphinyl]acetyl]-L-proline, Fosinopril impurity A, (2S,4S)-4-cyclohexyl-1-[[(R)-hydroxy(4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylic acid, USP Fosinopril Related Compound A, SQ-27519, (4S)-4-cyclohexyl-[(4-phenylbutyl)phosphinyl]acetyl-L-proline, Fosinopril Related Compound A, Fosinopril Sodium Impurity A, Fosinopril Impurity A
InChI formel InChI=1S/C23H34NO5P/c25-22(17-30(28,29)14-8-7-11-18-9-3-1-4-10-18)24-16-20(15-21(24)23(26)27)19-12-5-2-6-13-19/h1,3-4,9-10,19-21H,2,5-8,11-17H2,(H,26,27)(H,28,29)/t20-,21+/m1/s1
SMIL OC(=O)[C@@H]1C[C@H](CN1C(=O)CP(=O)(O)CCCCc2ccccc2)C3CCCCC3
IUPAC navn (2S,4S)-4-cyclohexyl-1-[2-[hydroxy(4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylic acid
Molekylvægt (g/mol) 435.493
Mængde 25 mg
Formel vægt 435.217 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.