Få mere at vide
(4S)-2-[Carboxy[[[3-(2-chloro-6-fluorophenyl)-5-methylisoxazol-4-yl]carbonyl]amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Disodium Salt (Penicilloic Acids of Flucloxacillin Disodium Salt), Mikromol™
Artikelnummer. 17982886
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
17982886 50 mg
1 options
Artikelnummer. 17982886 Leverandør Mikromol Leverandørnr. MM0489.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(4S)-2-[Carboxy[[[3-(2-chloro-6-fluorophenyl)-5-methylisoxazol-4-yl]carbonyl]amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Disodium Salt (Penicilloic Acids of Flucloxacillin Disodium Salt)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS,4S)-2-[(((3-(2-Chloro-6-fluorophenyl)-5-methylisoxazol-4-yl)carbonyl)amino)methyl]-5,5-dimethyl-thiazolidine-4-carboxylic Acid
CAS 1276016-89-7
Analyt- eller komponentnavne (2RS,4S)-2-[(((3-(2-Chloro-6-fluorophenyl)-5-methylisoxazol-4-yl)carbonyl)amino)methyl]-5,5-dimethyl-thiazolidine-4-carboxylic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C18 H19 Cl F N3 O4 S
Certificeringer/compliance ISO 9001
Synonym Flucloxacillin Sodium Imp. B (EP), Flucloxacillin Imp. B (EP), (2RS,4S)-2-[[[[3-(2-Chloro-6-fluorophenyl)-5-methylisoxazol-4-yl]carbonyl]amino]methyl]-5,5-dimethyl-thiazolidine-4-carboxylic Acid, Penilloic Acids of Flucloxacillin, Flucloxacillin Sodium Impurity B, Flucloxacillin Impurity B
InChI formel InChI=1S/C17H20N2S.ClH/c1-13(12-18(2)3)19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H
SMIL Cc1onc(c1C(=O)NCC2N[C@@H](C(=O)O)C(C)(C)S2)c3c(F)cccc3Cl
IUPAC navn (4S)-2-[[[3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]methyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid
Molekylvægt (g/mol) 427.88
Mængde 50 mg
Formel vægt 427.0769 g/mol