Få mere at vide
(4S)-2-[Carboxy[[[3-(2-chloro-6-fluorophenyl)-5-methylisoxazol-4-yl]carbonyl]amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Disodium Salt, Mikromol™
Artikelnummer. 30051276
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051276 25 mg
17942516 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051276 Leverandør Mikromol Leverandørnr. MM0489.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(4S)-2-[Carboxy[[[3-(2-chloro-6-fluorophenyl)-5-methylisoxazol-4-yl]carbonyl]amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Disodium Salt
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4S)-2-[Carboxy[((3-(2-chloro-6-fluorophenyl)-5-methylisoxazol-4-yl)carbonyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Disodium Salt
CAS 68728-50-7
Analyt- eller komponentnavne (4S)-2-[Carboxy[((3-(2-chloro-6-fluorophenyl)-5-methylisoxazol-4-yl)carbonyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Disodium Salt
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C19 H17 Cl F N3 O6 S . 2 Na
Certificeringer/compliance ISO 9001
Synonym Flucloxacillin Sodium Imp. A (EP), Flucloxacillin Imp. A (EP), (4S)-2-[Carboxy[[[3-(2-chloro-6-fluorophenyl)-5-methylisoxazol-4-yl]carbonyl]amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Disodium Salt, Flucloxacillin Sodium Imp. A (EP) as Disodium Salt, Penicilloic Acids of Flucloxacillin Disodium Salt, Flucloxacillin Sodium Impurity A as Disodium Salt, Flucloxacillin Impurity A as Disodium Salt
InChI formel InChI=1S/C19H19ClFN3O6S.2Na/c1-7-10(12(24-30-7)11-8(20)5-4-6-9(11)21)15(25)22-13(17(26)27)16-23-14(18(28)29)19(2,3)31-16;;/h4-6,13-14,16,23H,1-3H3,(H,22,25)(H,26,27)(H,28,29);;/q;2*+1/p-2/t13?,14-,16?;;/m0../s1
SMIL [Na+].[Na+].Cc1onc(c1C(=O)NC(C2N[C@@H](C(=O)[O-])C(C)(C)S2)C(=O)[O-])c3c(F)cccc3Cl
IUPAC navn disodium;(4S)-2-[carboxylato-[[3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]methyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylate
Molekylvægt (g/mol) 515.85
Mængde 25 mg
Formel vægt 515.0306 g/mol