Få mere at vide
(4S)-2-(3,6-Dioxo-5-phenylpiperazin-2-yl)-5,5-dimethylthiazolidine-4-carboxylic Acid (Diketopiperazines ofAmpicillin), Mikromol™
Artikelnummer. 30460432
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460432 4 x 25 mg
30051514 25 mg
17922506 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30460432 Leverandør Mikromol Leverandørnr. MM0253.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(4S)-2-(3,6-Dioxo-5-phenylpiperazin-2-yl)-5,5-dimethylthiazolidine-4-carboxylic Acid (Diketopiperazines ofAmpicillin), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4S)-2-(3,6-Dioxo-5-phenylpiperazin-2-yl)-5,5-dimethylthiazolidine-4-carboxylic Acid (Diketopiperazines ofAmpicillin)
CAS 49841-96-5
Analyt- eller komponentnavne (4S)-2-(3,6-Dioxo-5-phenylpiperazin-2-yl)-5,5-dimethylthiazolidine-4-carboxylic Acid (Diketopiperazines ofAmpicillin)
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C16 H19 N3 O4 S
Certificeringer/compliance ISO 9001
Synonym Ampicillin Sodium Imp. C (EP), Ampicillin Imp. C (EP), (4S)-2-(3,6-Dioxo-5-phenylpiperazin-2-yl)-5,5-dimethylthiazolidine-4-carboxylic Acid, Diketopiperazines ofAmpicillin, Ampicillin Sodium Impurity C, Ampicillin Impurity C
InChI formel InChI=1S/C16H19N3O4S/c1-16(2)11(15(22)23)19-14(24-16)10-13(21)17-9(12(20)18-10)8-6-4-3-5-7-8/h3-7,9-11,14,19H,1-2H3,(H,17,21)(H,18,20)(H,22,23)/t9?,10?,11-,14?/m0/s1
Løsningstype Neat
SMIL CC1(C)SC(N[C@H]1C(=O)O)C2NC(=O)C(NC2=O)c3ccccc3
IUPAC navn (4S)-2-(3,6-dioxo-5-phenylpiperazin-2-yl)-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid
Molekylvægt (g/mol) 349.4
Mængde 4 x 25 mg
Formel vægt 349.1096 g/mol