Få mere at vide
(4S)-2-[[[(2R)-2-Amino-2-phenylacetyl]amino]carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt (Penicilloic Acids of Ampicillin Sodium Salt), Mikromol™
Artikelnummer. 30462536
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462536 4 x 25 mg
30052606 25 mg
17934186 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462536 Leverandør Mikromol Leverandørnr. MM0253.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(4S)-2-[[[(2R)-2-Amino-2-phenylacetyl]amino]carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt (Penicilloic Acids of Ampicillin Sodium Salt), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Penicilloic Acids of Ampicillin Sodium Salt
CAS 1642559-70-3
Analyt- eller komponentnavne Penicilloic Acids of Ampicillin Sodium Salt
Forsendelsestilstand Room Temperature
Molekylær formel C16 H20 N3 O5 S . Na
Certificeringer/compliance ISO 9001
Synonym Ampicillin Sodium Imp. D (EP), Ampicillin Imp. D (EP), (4S)-2-[[[(2R)-2-Amino-2-phenylacetyl]amino]carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt, Ampicillin Sodium Imp. D (EP) as Sodium Salt, Penicilloic Acids of Ampicillin Sodium Salt, Ampicillin Sodium Impurity D as Sodium Salt, Ampicillin Impurity D as Sodium Salt
InChI formel InChI=1S/C25H30INO3.ClH/c1-4-7-11-23-24(19-10-8-9-12-21(19)30-23)25(28)18-13-14-22(20(26)17-18)29-16-15-27(5-2)6-3;/h8-10,12-14,17H,4-7,11,15-16H2,1-3H3;1H
Løsningstype Neat
SMIL [Na+].CC1(C)SC(N[C@H]1C(=O)[O-])C(NC(=O)[C@H](N)c2ccccc2)C(=O)O
IUPAC navn sodium;(4S)-2-[[[(2R)-2-amino-2-phenylacetyl]amino]-carboxymethyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylate
Molekylvægt (g/mol) 389.4
Mængde 4 x 25 mg
Formel vægt 389.1021 g/mol