Få mere at vide
(4S)-2-[[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt (Penicilloic Acids of Amoxicillin Sodium Salt), Mikromol™
Beskrivelse
(4S)-2-[[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt (Penicilloic Acids of Amoxicillin Sodium Salt)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (4S)-2-[([(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino)carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt (Penicilloic Acids of Amoxicillin Sodium Salt) |
| CAS | 68728-47-2 |
| Analyt- eller komponentnavne | (4S)-2-[([(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino)carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt (Penicilloic Acids of Amoxicillin Sodium Salt) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H20 N3 O6 S . Na |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Amoxicillin Trihydrate Imp. D (EP), Amoxicillin Imp. D (EP), Amoxicillin USP Related Compound D, Amoxicillin USP RC D, (4S)-2-[[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt, 2-Thiazolidineacetic acid, alpha-[[amino(4-hydroxyphenyl)acetyl]amino]-4-carboxy-5,5-dimethyl-, monosodium salt (9CI), Amoxicillin Trihydrate Imp. D (EP) as Sodium Salt, Penicilloic Acids of Amoxicillin Sodium Salt, Amoxicillin Related Compound D as Sodium Salt, Amoxicillin Trihydrate Impurity D as Sodium Salt, Amoxicillin Impurity D as Sodium Salt |
| InChI formel | InChI=1S/C16H21N3O6S.Na/c1-16(2)11(15(24)25)19-13(26-16)10(14(22)23)18-12(21)9(17)7-3-5-8(20)6-4-7;/h3-6,9-11,13,19-20H,17H2,1-2H3,(H,18,21)(H,22,23)(H,24,25);/q;+1/p-1/t9-,10?,11+,13?;/m1./s1 |
| SMIL | [Na+].CC1(C)SC(N[C@H]1C(=O)[O-])C(NC(=O)[C@H](N)c2ccc(O)cc2)C(=O)O |
| IUPAC navn | sodium;(4S)-2-[[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-carboxymethyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylate |
| Vis mere |