Få mere at vide
(4S)-2-[[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt (Penicilloic Acids of Amoxicillin Sodium Salt), Mikromol™
Artikelnummer. 17218509
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17218509 100 mg
30051686 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17218509 Leverandør Mikromol Leverandørnr. MM0232.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(4S)-2-[[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt (Penicilloic Acids of Amoxicillin Sodium Salt)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4S)-2-[([(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino)carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt (Penicilloic Acids of Amoxicillin Sodium Salt)
CAS 68728-47-2
Analyt- eller komponentnavne (4S)-2-[([(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino)carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt (Penicilloic Acids of Amoxicillin Sodium Salt)
Forsendelsestilstand Room Temperature
Molekylær formel C16 H20 N3 O6 S . Na
Certificeringer/compliance ISO 9001
Synonym Amoxicillin Trihydrate Imp. D (EP), Amoxicillin Imp. D (EP), Amoxicillin USP Related Compound D, Amoxicillin USP RC D, (4S)-2-[[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]carboxymethyl]-5,5-dimethylthiazolidine-4-carboxylic Acid Sodium Salt, 2-Thiazolidineacetic acid, alpha-[[amino(4-hydroxyphenyl)acetyl]amino]-4-carboxy-5,5-dimethyl-, monosodium salt (9CI), Amoxicillin Trihydrate Imp. D (EP) as Sodium Salt, Penicilloic Acids of Amoxicillin Sodium Salt, Amoxicillin Related Compound D as Sodium Salt, Amoxicillin Trihydrate Impurity D as Sodium Salt, Amoxicillin Impurity D as Sodium Salt
InChI formel InChI=1S/C16H21N3O6S.Na/c1-16(2)11(15(24)25)19-13(26-16)10(14(22)23)18-12(21)9(17)7-3-5-8(20)6-4-7;/h3-6,9-11,13,19-20H,17H2,1-2H3,(H,18,21)(H,22,23)(H,24,25);/q;+1/p-1/t9-,10?,11+,13?;/m1./s1
SMIL [Na+].CC1(C)SC(N[C@H]1C(=O)[O-])C(NC(=O)[C@H](N)c2ccc(O)cc2)C(=O)O
IUPAC navn sodium;(4S)-2-[[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-carboxymethyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylate
Molekylvægt (g/mol) 405.4
Mængde 100 mg
Formel vægt 405.0971 g/mol